Learn More
Medchemexpress LLC Dotap (chloride) | 132172-61-3 | 99.9% | 698.54 | 250 MG
SDP
Catalog No. 5000439486
Encompass_Preferred
DOTAP chloride is a useful and effective cationic lipid for transient and stable transfection of DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides) without the use of a helper lipid. It is intended for research use only and not for sale to patients.
  • Useful and effective cationic lipid for transient and stable transfection of DNA and modified nucleic acids.
  • Does not require the use of a helper lipid for transfection.
Catalog No. 50-004-39486 Supplier Medchemexpress LLC Supplier No. 250MGHY112754AGL
DOTAP chloride is a useful and effective cationic lipid for transient and stable transfection of DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides) without the use of a helper lipid. It is intended for research use only and not for sale to patients.
  • Useful and effective cationic lipid for transient and stable transfection of DNA and modified nucleic acids.
  • Does not require the use of a helper lipid for transfection.