Learn More
Kyfora Bio DOTAP CHLORIDE 500 MG
Catalog No. 502559851
Encompass_Preferred
DOTAP Chloride (N-[1-(2,3-Dioleoyloxy)propyl]-N,N,N-trimethylammonium chloride) is a cationic lipid widely used for in vitro and in vivo delivery of DNA, RNA, and other negatively charged molecules. DOTAP encapsulates the nucleic acids to form a stable complex spontaneously by means of electrostatic interaction. It can also be used to form lipid nanoparticles (LNPs) for mRNA vaccine delivery and unilamellar liposomes for gene delivery. This product is for research use only.Key Application(s): Gene delivery, mRNA delivery
Catalog No. 50-255-9851 Supplier Kyfora Bio Supplier No. 14475500
DOTAP Chloride (N-[1-(2,3-Dioleoyloxy)propyl]-N,N,N-trimethylammonium chloride) is a cationic lipid widely used for in vitro and in vivo delivery of DNA, RNA, and other negatively charged molecules. DOTAP encapsulates the nucleic acids to form a stable complex spontaneously by means of electrostatic interaction. It can also be used to form lipid nanoparticles (LNPs) for mRNA vaccine delivery and unilamellar liposomes for gene delivery. This product is for research use only.Key Application(s): Gene delivery, mRNA delivery