Learn More
Medchemexpress LLC DOTAP chloride (GMP Like) | 132172-61-3 | 698.54 | 50 MG
SDP
Catalog No. 5000438579
Encompass_Preferred
DOTAP chloride (GMP Like) is a GMP Like class of DOTAP chloride that serves as an auxiliary reagent for cell therapy manufacture. It is a cationic lipid known for its good membrane fusion ability and biocompatibility. DOTAP chloride (GMP Like) can function as an excipient for transient and stable transfection of DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides) without requiring a helper lipid.
  • Gmp like class DOTAP chloride
  • Auxiliary reagent for cell therapy manufacture
  • Cationic lipid
  • Good membrane fusion ability
  • Biocompatibility
  • Can be used as an excipient for transient and stable transfection of DNA and modified nucleic acids without helper lipid
Catalog No. 50-004-38579 Supplier Medchemexpress LLC Supplier No. HY112754AGL50MG
DOTAP chloride (GMP Like) is a GMP Like class of DOTAP chloride that serves as an auxiliary reagent for cell therapy manufacture. It is a cationic lipid known for its good membrane fusion ability and biocompatibility. DOTAP chloride (GMP Like) can function as an excipient for transient and stable transfection of DNA (plasmids, bacmids) and modified nucleic acids (antisense oligonucleotides) without requiring a helper lipid.
  • Gmp like class DOTAP chloride
  • Auxiliary reagent for cell therapy manufacture
  • Cationic lipid
  • Good membrane fusion ability
  • Biocompatibility
  • Can be used as an excipient for transient and stable transfection of DNA and modified nucleic acids without helper lipid