Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DOTAP-d9 iodide | 00-00-0 | 100.0% | 799.05 | C42H71D9INO4 | 5 MG
SDP

Catalog No. 5000222304
Encompass_Preferred

DOTAP-d9 iodide is a deuterium-labeled cationic lipid (iodide salt) supplied as a solid for use as a stable isotope tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is provided in small research quantities with high stated purity and specified storage conditions.

  • Deuterium-labeled analog suitable for tracer studies and isotopic tracing.
  • Can be used as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity (99.97%) for analytical applications.
  • Solid, white to off-white appearance for easy handling and identification.
  • Molecular weight 799.05 and formula C42H71D9INO4 for accurate mass calculations.
  • Recommended storage: -20 °C sealed; in solvent stability: -80 °C (6 months), -20 °C (1 month).

Catalog No. 50-002-22304 Supplier Medchemexpress LLC Supplier No. HY143950S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DOTAP-d9 iodide is a deuterium-labeled cationic lipid (iodide salt) supplied as a solid for use as a stable isotope tracer or internal standard in quantitative NMR, GC-MS, and LC-MS analyses. It is provided in small research quantities with high stated purity and specified storage conditions.

  • Deuterium-labeled analog suitable for tracer studies and isotopic tracing.
  • Can be used as an internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity (99.97%) for analytical applications.
  • Solid, white to off-white appearance for easy handling and identification.
  • Molecular weight 799.05 and formula C42H71D9INO4 for accurate mass calculations.
  • Recommended storage: -20 °C sealed; in solvent stability: -80 °C (6 months), -20 °C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.