Learn More
Kyfora Bio DOTAP METHYL SULFATE 25 MG
Supplier:  Kyfora Bio 14475MS25
Encompass_Preferred
DOTAP MS (N-[1-(2,3-Dioleoyloxy)propyl]-N,N,N-trimethylammonium methyl-sulfate) is a proven reagent for the highly efficient liposomal transfection of DNA, RNA, oligonucleotides, other negatively charged biomolecules. Several other techniques of introducing genetic material into cells may suffer from inadequate reproducibility, cytotoxicity, inconvenience of process, and low transfection efficiency. DOTAP avoids these drawbacks and works by encapsulating the negatively charged DNA within a spontaneously formed liposomal vesicle. This DOTAP/DNA complex can then fuse with the cell membrane and insert DNA directly into the cytoplasm. This mechanism of action ensures transfection occurs efficiently and with little to no cytotoxicity.
Catalog No. 50-255-9849