Learn More
Medchemexpress LLC DOTATATE acetate 10mg | 177943-89-4 | 10MG
SDP
Catalog No. 5000243720
Encompass_Preferred
DOTATATE acetate is a DOTA-conjugated peptide DOTATATE acetate can be labelled with radionuclides for positron emission tomography (PET) imaging and peptide receptor radionuclide research (PRRT)[1 [2 [3 [4 DOTATATE (acetate) can be used for the synthesis/research of Radionuclide-Drug Conjugates (RDCs)
Catalog No. 50-002-43720 Supplier Medchemexpress LLC Supplier No. HY106244A10MG
DOTATATE acetate is a DOTA-conjugated peptide DOTATATE acetate can be labelled with radionuclides for positron emission tomography (PET) imaging and peptide receptor radionuclide research (PRRT)[1 [2 [3 [4 DOTATATE (acetate) can be used for the synthesis/research of Radionuclide-Drug Conjugates (RDCs)