Learn More
Medchemexpress LLC Dotatate (acetate) | 177943-89-4 | >98.0% | 1435.6 g/mol | C65H90N14O19S2 · xC2H4O2 | 100 MG
SDP
Catalog No. 5000243834
Encompass_Preferred
DOTATATE (acetate) is a DOTA-conjugated peptide used in research for positron emission tomography (PET) imaging and peptide receptor radionuclide therapy. It functions as a precursor for radiolabeling with diagnostic and therapeutic radionuclides and for synthesis of radionuclide-drug conjugates, supplied as an acetate salt for laboratory use.
  • High purity (>98%).
  • Suitable for radiolabeling with diagnostic and therapeutic radionuclides.
  • Provided with supporting documents (datasheet, SDS, COA).
  • Available in multiple package sizes including 100 MG.
  • Peptide format designed for chelation and stable conjugation.
Catalog No. 50-002-43834 Supplier Medchemexpress LLC Supplier No. HY106244A100MG
DOTATATE (acetate) is a DOTA-conjugated peptide used in research for positron emission tomography (PET) imaging and peptide receptor radionuclide therapy. It functions as a precursor for radiolabeling with diagnostic and therapeutic radionuclides and for synthesis of radionuclide-drug conjugates, supplied as an acetate salt for laboratory use.
  • High purity (>98%).
  • Suitable for radiolabeling with diagnostic and therapeutic radionuclides.
  • Provided with supporting documents (datasheet, SDS, COA).
  • Available in multiple package sizes including 100 MG.
  • Peptide format designed for chelation and stable conjugation.