Learn More
Medchemexpress LLC Dotatate acetate | 177943-89-4 | MFCD09039058 | ≥98.0% | 1435.6 g/mol | C67H94N14O21S2 | 25 MG
SDP
Catalog No. 5000241837
Encompass_Preferred
DOTATATE acetate is a DOTA-conjugated somatostatin analogue peptide used in radiochemistry research. The peptide acts as a bifunctional chelator-conjugated ligand for radiolabeling with metallic radionuclides for PET imaging and for peptide receptor radionuclide therapy research, targeting somatostatin receptor-expressing tumors such as neuroendocrine tumors.
  • Suitable for radiolabeling with DOTA-compatible radionuclides for PET and therapy
  • High chemical purity for reproducible conjugation and labeling
  • Targets somatostatin receptors for receptor-specific imaging and therapy studies
  • Available as an acetate salt and supplied lyophilized for stability
  • Packaged in small milligram quantities suitable for preclinical research
Catalog No. 50-002-41837 Supplier Medchemexpress LLC Supplier No. HY106244A25MG
DOTATATE acetate is a DOTA-conjugated somatostatin analogue peptide used in radiochemistry research. The peptide acts as a bifunctional chelator-conjugated ligand for radiolabeling with metallic radionuclides for PET imaging and for peptide receptor radionuclide therapy research, targeting somatostatin receptor-expressing tumors such as neuroendocrine tumors.
  • Suitable for radiolabeling with DOTA-compatible radionuclides for PET and therapy
  • High chemical purity for reproducible conjugation and labeling
  • Targets somatostatin receptors for receptor-specific imaging and therapy studies
  • Available as an acetate salt and supplied lyophilized for stability
  • Packaged in small milligram quantities suitable for preclinical research