Learn More
TARGETMOL CHEMICALS INC DOTATATE acetate 5MG
SDP
Catalog No. 502877120
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DOTATATE acetate is a DOTA-binding peptide, a growth inhibitor analog, and a novel PET tracer involved in the assessment of functional imaging of highly differentiated neuroendocrine tumors (NETs). Purity 98.7%
Catalog No. 50-287-7120 Supplier TARGETMOL CHEMICALS INC Supplier No. T391935MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. DOTATATE acetate is a DOTA-binding peptide, a growth inhibitor analog, and a novel PET tracer involved in the assessment of functional imaging of highly differentiated neuroendocrine tumors (NETs). Purity 98.7%