Learn More
Medchemexpress LLC HY-106244A 5mg , DOTATATE (acetate) CAS:177943-89-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106244A5MG
Encompass_Preferred
HY-106244A 5mg DOTATATE (acetate) CAS: 177943-89-4 DOTATATE acetate is a DOTA-conjugated peptide. DOTATATE acetate can be labelled with radionuclides for positron emission tomography (PET) imaging and peptide receptor radionuclide therapy (PRRT)[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0482