Learn More
Cayman Chemical DOTatTEacetate 10mg
SDP
Catalog No. 502944658
Encompass_Preferred
A synthetic peptide; has been conjugated to various radioligands for use in scintigraphy and PET/CT imaging of SST-positive human tumor slices
Catalog No. 50-294-4658 Supplier Cayman Chemical Supplier No. 4105410 MG
A synthetic peptide; has been conjugated to various radioligands for use in scintigraphy and PET/CT imaging of SST-positive human tumor slices