Learn More
Cayman Chemical DOTATATE (acetate), Purity: ≥98%, CAS: 177943-89-4, 5 mg
SDP
Catalog No. 502978555
Encompass_Preferred
A synthetic peptide; has been conjugated to various radioligands for use in scintigraphy and PET/CT imaging of SST-positive human tumor slices
Catalog No. 50-297-8555 Supplier Cayman Chemical Supplier No. 410545 MG
A synthetic peptide; has been conjugated to various radioligands for use in scintigraphy and PET/CT imaging of SST-positive human tumor slices