Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dotetracontane, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015316 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5316 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5316 Supplier MilliporeSigma Supelco Supplier No. 44250100MG
Add to Cart
Add to Cart

Dotetracontane is classified under the high molecular weight non-polar hydrocarbon group of compounds.

Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 7098-20-6
Physical Form Neat
Linear Formula CH3(CH2)40 CH3
MDL Number MFCD00665974
Melting Point 83°C to 86°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 591.13
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.