Learn More
Medchemexpress LLC HY-109031 10mg Medchemexpress, Dotinurad CAS:1285572-51-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10903110MG
Encompass_Preferred
Medchemexpress, HY-109031 10mg Dotinurad CAS:1285572-51-1 Dotinurad is a potent uricosuric agent, with an IC50 of 3.6 µM for uric acid. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8652