Learn More
Apexbio Technology LLC DOTMA 104872-42-6 250mg
SDP
Supplier:  Apexbio Technology LLC M2003250
Encompass_Preferred
DOTMA (CAS 104872-42-6) is a tetramethylated DOTA analogue structurally classified as a cationic lipid It imparts a positive surface charge to liposomes thus enhancing electrostatic interactions with negatively charged cellular membranes This property facilitates cellular uptake enabling its use as a non-viral vector for nucleic acid delivery including siRNA microRNA and oligonucleotides DOTMA is widely utilized in both in vitro and in vivo gene transfection studies contributing to research in gene therapy and nucleic acid-based therapeutics
Catalog No. 50-000-97030