Learn More
Medchemexpress LLC DOTMA | 104872-42-6 | 99.91% | 670.57 | 200 MG
SDP
Catalog No. 5000349254
Encompass_Preferred
DOTMA is a cationic lipid used as a non-viral vector for gene therapy. It is a component of liposomes for encapsulating siRNA, microRNA, and oligonucleotides, and for in vitro gene transfection. DOTMA promotes effective interaction between liposomes and cell membranes by inducing positive charge on the liposomes. It has demonstrated good gene transfection effects both in vitro and in vivo.
  • Cationic lipid
  • Non-viral vector for gene therapy
  • Component of liposomes for encapsulating siRNA, microRNA, and oligonucleotides
  • Used for in vitro gene transfection
  • Promotes effective interaction between liposomes and cell membranes
  • Induces positive charge on liposomes
  • Good gene transfection effects both in vitro and in vivo
  • High intravenous transfection activity in CD-1 mouse
Catalog No. 50-003-49254 Supplier Medchemexpress LLC Supplier No. HY139200200MG
DOTMA is a cationic lipid used as a non-viral vector for gene therapy. It is a component of liposomes for encapsulating siRNA, microRNA, and oligonucleotides, and for in vitro gene transfection. DOTMA promotes effective interaction between liposomes and cell membranes by inducing positive charge on the liposomes. It has demonstrated good gene transfection effects both in vitro and in vivo.
  • Cationic lipid
  • Non-viral vector for gene therapy
  • Component of liposomes for encapsulating siRNA, microRNA, and oligonucleotides
  • Used for in vitro gene transfection
  • Promotes effective interaction between liposomes and cell membranes
  • Induces positive charge on liposomes
  • Good gene transfection effects both in vitro and in vivo
  • High intravenous transfection activity in CD-1 mouse