Learn More
Medchemexpress LLC HY-139200 50mg , DOTMA CAS:104872-42-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13920050MG
Encompass_Preferred
HY-139200 50mg DOTMA CAS: 104872-42-6 DOTMA, as a tetra-methylated DOTA analogue, is a cationic lipid and can be used as a non-viral vector for gene therapy. It has been used as a component in liposomes that can be used to encapsulate siRNA, microRNAs, and oligonucleotides and for gene transfection in vitro. It exhibits effective in vitro and in vivo gene transfection. DOTMA induces a positive charge on the liposomes and thus promotes efficient liposome- cell membrane interaction. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0824