Learn More
Medchemexpress LLC Dotriacontane-d66 | 62369-68-0 | 99.09% | 517.27 | 50 MG
SDP
Catalog No. 5000426103
Encompass_Preferred
Dotriacontane-d66 is the deuterium labeled Dotriacontane. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-26103 Supplier Medchemexpress LLC Supplier No. HYN9600S50MG
Dotriacontane-d66 is the deuterium labeled Dotriacontane. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS