Learn More
Medchemexpress LLC Dotriacontane-d66 | 62369-68-0 | 99.1% | 517.27 | 10 MG
SDP
Catalog No. 5000426576
Encompass_Preferred
Dotriacontane-d66 is a deuterium-labeled form of Dotriacontane, used primarily as a tracer and internal standard. It plays a crucial role in quantitative analysis methods such as NMR, GC-MS, and LC-MS, leveraging the unique properties of stable heavy isotopes. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes into drug molecules
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-26576 Supplier Medchemexpress LLC Supplier No. HYN9600S10MG
Dotriacontane-d66 is a deuterium-labeled form of Dotriacontane, used primarily as a tracer and internal standard. It plays a crucial role in quantitative analysis methods such as NMR, GC-MS, and LC-MS, leveraging the unique properties of stable heavy isotopes. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Incorporates stable heavy isotopes into drug molecules
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs