Learn More
Medchemexpress LLC Dotriacontane-d66 | 62369-68-0 | 99.1% | 517.27 | 100 MG
SDP
Catalog No. 5000425415
Encompass_Preferred
Dotriacontane-d66 is a deuterium labeled Dotriacontane. It can be used as a tracer and as an internal standard for quantitative analysis. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-25415 Supplier Medchemexpress LLC Supplier No. HYN9600S100MG
Dotriacontane-d66 is a deuterium labeled Dotriacontane. It can be used as a tracer and as an internal standard for quantitative analysis. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs