Learn More
Medchemexpress LLC Dotriacontane-d66 | 62369-68-0 | 99.1% | 517.27 | 25 MG
SDP
Catalog No. 5000426951
Encompass_Preferred
Dotriacontane-d66 is a deuterium-labeled Dotriacontane, used for research purposes as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-26951 Supplier Medchemexpress LLC Supplier No. 25MGHYN9600S
Add to Cart
Add to Cart
Dotriacontane-d66 is a deuterium-labeled Dotriacontane, used for research purposes as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS