Learn More
Cayman Chemical Dotriacontanoic Acid methyl ester, Purity: ≥98%, CAS: 41755-79-7, 50 mg
SDP
Catalog No. 502934944
Encompass_Preferred
A naturally occurring fatty acid methyl ester; has been found in the cuticular wax of P. abies needles; has been found in sediment samples from the Harney River in Florida and Lake Kivu in the East African rift valley
Catalog No. 50-293-4944 Supplier Cayman Chemical Supplier No. 2672850 MG
A naturally occurring fatty acid methyl ester; has been found in the cuticular wax of P. abies needles; has been found in sediment samples from the Harney River in Florida and Lake Kivu in the East African rift valley