Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Dotted Grip Scalpel Handle, No.8

Catalog No. 21124579 Shop All Surgical Design Products
Change view
Click to view available options
Color:
Gold
Silver
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-124-579 Gold
21-124-580 -
21-124-581 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-124-579 Supplier Surgical Design™ Supplier No. 2728NPGD
Add to Cart
Add to Cart

Dotted grip scalpel handle offers control and comfort with a textured anti-slip grip for precision handling.

  • Made from premium German stainless steel for precision, durability, and corrosion resistance
  • Dotted anti-slip grip ensures steady handling and maximum control, even in wet conditions
  • Compatible with No. 60-70 surgical blades, ideal for fine and controlled incisions
  • Available in three high-polish finishes — Silver, Gold, and Multicolor Titanium
  • Lightweight and balanced for improved tactile feedback during dissection and surgery
  • Ergonomic profile reduces hand strain and enhances stability during extended use
  • Mirror and titanium coatings resist rust and staining while adding visual appeal
  • Autoclavable and reusable, maintaining performance over repeated sterilization cycles
  • Crafted for professional surgical and laboratory use
  • Ideal for surgical, anatomical, and veterinary dissection
  • Suitable for medical training, laboratory research, and precision cutting

Specifications

Product Type Scalpel Handle
Size No.8
For Use With (Equipment) 60 to 70 Scalpel Blades
Disposable Reusable
Sterility Non-sterile
Certifications/Compliance ISO/GMP
Autoclavable Autoclavable
Handle Material Stainless Steel
Color Gold
Graduated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.