Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Dotted Grip Scalpel Handle

Catalog No. 21124582 Shop All Surgical Design Products
Change view
Click to view available options
Size:
No.3
No.4
No.7
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-124-582 No.7
21-124-577 No.3
21-124-578 No.4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-124-582 Supplier Surgical Design™ Supplier No. 272NP
Only null left
Add to Cart
Add to Cart

Dotted grip scalpel handle ensures firm, slip-free control and precision during dissection.

  • Made from high-quality surgical-grade stainless steel for strength, balance, and long-lasting performance
  • Dotted anti-slip grip ensures maximum control and stability during fine surgical or laboratory dissection
  • Available in Sizes No. 3, 4, and 7, compatible with a wide range of standard surgical blades
  • Ergonomic handle design minimizes hand fatigue and provides superior tactile feedback
  • Precision ruler markings (on select models) aid in accurate measurement and depth control
  • Corrosion-resistant finish for easy sterilization and long-term use in demanding environments
  • Lightweight and balanced for comfort during extended procedures
  • Autoclavable and reusable, ensuring professional-grade hygiene standards
  • Designed for precision incisions, tissue separation, and educational dissection
  • Ideal for surgical, laboratory, anatomical, and veterinary use
  • Commonly used in dissection, tissue preparation, and surgical training
  • Suitable for operating rooms, medical schools, and pathology labs

Specifications

Product Type Scalpel Handle
Size No.7
Length (English) Handle 6.5 in.
Length (Metric) Handle 16.5 cm
For Use With (Equipment) 10 Series Scalpel Blades
Disposable Reusable
Sterility Non-sterile
Certifications/Compliance ISO/GMP
Autoclavable Autoclavable
Handle Material Stainless Steel
Color Silver
Graduated No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.