Learn More
Roboz Surgical DOU SIDE NYLON BRISTLE BRU 3/P
SDP
Catalog No. NC0513024
Encompass
Double sided nylon brush. Perfect for cleaning blades, hinges, and serrated areas of all surgical instruments. 7in length. 5mm and 10mm bristles.
Catalog No. NC0513024 Supplier Roboz Surgical Supplier No. IC1001
Double sided nylon brush. Perfect for cleaning blades, hinges, and serrated areas of all surgical instruments. 7in length. 5mm and 10mm bristles.