Learn More
Ted Pella Inc Double 90º SEM Mount, Ø25mm
SDP
Catalog No. NC1067284
Encompass
Double 90º SEM Mount Ø25mm, pin leg fits into FEI, Tescan, ZEISS (also for Philips, Leo, Cambridge, AMRAY, Leica, CamScan, ETEC) SEMs. Aluminum, grooved edge, 9.5mm pin height.
Catalog No. NC1067284 Supplier Ted Pella Inc Supplier No. 16354
Double 90º SEM Mount Ø25mm, pin leg fits into FEI, Tescan, ZEISS (also for Philips, Leo, Cambridge, AMRAY, Leica, CamScan, ETEC) SEMs. Aluminum, grooved edge, 9.5mm pin height.