Learn More
Diversified Biotech Inc DOUBLE ALUM CHAMBER EA
Catalog No. 50589601
Encompass
Rack, Cooling Rack for 96 Well Plates, Gold Anodized Aluminum, Accommodates two 96 Well Plates, Keep Samples Cool Using an Ice Pan or Bucket, Direct Contact with Ice/Dry Ice/Liquid Nitrogen, 6.125 x 6.75 x 1.- Ideal Use 0.2ml PCR plates, EA
Catalog No. 50-589-601 Supplier Diversified Biotech Inc Supplier No. CHAM1020
Add to Cart
Add to Cart
Rack, Cooling Rack for 96 Well Plates, Gold Anodized Aluminum, Accommodates two 96 Well Plates, Keep Samples Cool Using an Ice Pan or Bucket, Direct Contact with Ice/Dry Ice/Liquid Nitrogen, 6.125 x 6.75 x 1.- Ideal Use 0.2ml PCR plates, EA