Learn More
Thermo Scientific™ Double Bagged Bottled Fluid Thioglycollate Medium (FTM)
Shop All Thermo Scientific ProductsDescription
Perform confident sterility and microbial limits testing with Thermo Scientific™ Remel™ Double Bagged Bottled Fluid Thioglycollate Medium (FTM) for testing according to USP, JP, and EU regulations and as a general purpose enrichment broth for cultivation and isolation of microorganisms. Double Bagged Bottled Fluid Thioglycollate Medium is ideal for microbiological testing that requires additional packaging for sterility protection between environmental class changes.
Specifications
Specifications
| Cap Type | Gold Metal Cap |
| Content And Storage | 2 to 25°C |
| Description | Double Bagged Bottled |
| Format | Dbl Bagged Wide Mouth Bottle |
| Product Type | Culture Medium |
| Quantity | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.