Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Double Bagged Bottled Tryptone Soya Broth (TSB)

Catalog No. R112996 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
Dbl Bagged Borosilicate Bottle
Dbl Bagged Wide Mouth Bottle
Cap Type:
PP plastic cap w/ Teflon
White Metal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Cap Type
R112996 Dbl Bagged Wide Mouth Bottle White Metal
R112912 Dbl Bagged Borosilicate Bottle PP plastic cap w/ Teflon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R112996 Supplier Thermo Scientific™ Supplier No. R112996
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform confident sterility testing with double bagged bottled media for added security between environmental class changes.

Perform confident sterility and microbial limits testing with Thermo Scientific™ Remel™ Double Bagged Bottled Tryptone Soya Broth (TSB) for testing according to USP, JP, and EU regulations and as a general purpose enrichment broth for cultivation and isolation of microorganisms. Double Bagged Bottled Tryptone Soya Broth (TSB) is ideal for microbiological testing that requires additional packaging for sterility protection between environmental class changes.

Add Remel Double Bagged Bottled Tryptone Soya Broth (TSB) to your sterility and microbial limits testing routine. Our range of validated solutions for the pharmaceutical microbiology laboratory support the production and quality assurance of sterile pharmaceuticals, vaccines, and biotechnology products.Complete Solutions, Complete Confidence:• Secure packaging: double bagged prior to autoclaving to preserve integrity of sterility media• Regulatory compliant: USP, JP, and EU harmonized media • Adaptable: a range of available fill volumes, media and bottle types to accommodate your sample sizes and performance testing needs• Quality guaranteed: Certificates of Analysis with every lot produced • Easy-to-use: simply ‘peel and pop’ bags for quick access to the media bottle• Multi-purpose media: suitable for general quality testing Customizable formats and media types are available. Please inquire.A highly nutritious versatile medium is recommended for general laboratory use. Due to the inclusion of both Tryptone and Soya Peptone, the medium will support a luxuriant growth of many fastidious organisms without the addition of serum.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Cap Type White Metal
Content And Storage 2 to 25°C
Description Double Bagged Bottled Tryptone Soya Broth (TSB )
Format Dbl Bagged Wide Mouth Bottle
Product Type Broth
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.