Learn More
Azzota Corp Double Beam Spectrophotometer, Scanning Function, 1.8Nm Bandwidth, 190-1100Nm, 320 X 240 Lcd. Auto Set Wl. Usb Connect. Displays/Saves 200 Standard Curves/Data. W/Kinetics Function, Pc Software
Catalog No. 501612951
Encompass_Preferred
Double Beam spectrophotometer, scanning function, 1.8nm bandwidth, 190-1100nm, 320 x 240 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. With Kinetics Function. PC software included.
Catalog No. 50-161-2951 Supplier Azzota Corp Supplier No. SE6100PC
Add to Cart
Add to Cart
Request a Quote
Double Beam spectrophotometer, scanning function, 1.8nm bandwidth, 190-1100nm, 320 x 240 LCD. Automatically set WL. USB connection. Can display and save 200 standard curves and 200 groups of data. With Kinetics Function. PC software included.