Learn More
MSC Vikan Replacement Cassette, Hygienic, 500 mm, White, Polypropylene/TPE
Supplier: MSC 45882230
This replacement cassette is designed for hygienic squeegees, ideal for environments where hygiene and food safety are critical, such as food manufacturing and food service. Featuring a unique double blade, it efficiently removes water and debris from various surfaces, including tiled and uneven floors. The easily removable design ensures improved cleanability, cost reduction, and environmental benefits, all within a durable construction that resists harsh chemicals.
- Purpose-built for critical hygiene and food safety environments.
- Unique double blade design offers balanced hygiene and efficiency.
- Easily removable cassette improves cleanability and reduces costs.
- Flexible blade effectively removes water and debris from various surfaces.
- Durable construction withstands harsh chemicals and daily use.
- Color-coded for hygienic zoning plans.