Learn More
MSC Vikan Replacement Cassette, Hygienic Squeegee Blade, 23.6 in, Green, Polypropylene/TPE
Catalog No. 502836423
Encompass
This green replacement squeegee blade for hygienic fixed and revolving neck squeegees is constructed from polypropylene and TPE rubber. Manufactured in Denmark, it measures 23.6 in L x 1 in W x 1.8 in H and weighs 0.375 lb.
  • Designed for critical hygiene areas like food manufacturing and retail.
  • Unique double blade balances hygiene and cleaning efficiency.
  • Easily removable replacement cassette improves cleanability and reduces costs.
  • Flexible blade follows surface contours for effective water and debris removal.
  • Durable construction withstands harsh chemicals and daily use.
  • Color-coded for use with hygienic zoning plans.
Catalog No. 50-283-6423 Supplier MSC Supplier No. 65343709
This green replacement squeegee blade for hygienic fixed and revolving neck squeegees is constructed from polypropylene and TPE rubber. Manufactured in Denmark, it measures 23.6 in L x 1 in W x 1.8 in H and weighs 0.375 lb.
  • Designed for critical hygiene areas like food manufacturing and retail.
  • Unique double blade balances hygiene and cleaning efficiency.
  • Easily removable replacement cassette improves cleanability and reduces costs.
  • Flexible blade follows surface contours for effective water and debris removal.
  • Durable construction withstands harsh chemicals and daily use.
  • Color-coded for use with hygienic zoning plans.