Learn More
MSC Vikan Hygienic Floor Squeegee, 500 mm, Polypropylene, White
Catalog No. 502835878
Encompass
This hygienic floor squeegee is designed for effective water removal in environments requiring high levels of hygiene. It can be used on various surfaces, including tiled and safety flooring. Its intelligent design combines maximum efficiency with high hygiene standards, offering the cleaning capacity of double-blade squeegees without compromising the hygiene of single-blade ultra-hygiene models.
  • Designed for areas requiring high levels of hygiene and effective water removal
  • Suitable for all types of surfaces, including tiled and safety flooring
  • Easily removable cassette for cleaning or replacement
  • Features a reinforced polypropylene head with specially blended rubber blades
  • Complies with EC 1935/2004 on food contact materials
  • Complies with FDA Regulation CFR 21
Catalog No. 50-283-5878 Supplier MSC Supplier No. 44926178
This hygienic floor squeegee is designed for effective water removal in environments requiring high levels of hygiene. It can be used on various surfaces, including tiled and safety flooring. Its intelligent design combines maximum efficiency with high hygiene standards, offering the cleaning capacity of double-blade squeegees without compromising the hygiene of single-blade ultra-hygiene models.
  • Designed for areas requiring high levels of hygiene and effective water removal
  • Suitable for all types of surfaces, including tiled and safety flooring
  • Easily removable cassette for cleaning or replacement
  • Features a reinforced polypropylene head with specially blended rubber blades
  • Complies with EC 1935/2004 on food contact materials
  • Complies with FDA Regulation CFR 21