Learn More
MSC Vikan Hygienic Floor Squeegee, 19.7 in, Polypropylene, TPE Rubber, Red
Catalog No. 502837166
Encompass
This hygienic floor squeegee is designed for environments demanding high levels of hygiene and effective water removal. It is suitable for use on various surfaces, including tiled and safety flooring, and features a removable cassette for easy cleaning or replacement.
  • Features a replacement cassette for easy maintenance.
  • Constructed from polypropylene and TPE rubber for durability.
  • Suitable for all types of surfaces, including tiled and safety flooring.
  • Complies with FDA Regulation CFR 21 and EU food contact material regulations.
  • Designed for areas requiring high levels of hygiene and effective water removal.
Catalog No. 50-283-7166 Supplier MSC Supplier No. 44926517
This hygienic floor squeegee is designed for environments demanding high levels of hygiene and effective water removal. It is suitable for use on various surfaces, including tiled and safety flooring, and features a removable cassette for easy cleaning or replacement.
  • Features a replacement cassette for easy maintenance.
  • Constructed from polypropylene and TPE rubber for durability.
  • Suitable for all types of surfaces, including tiled and safety flooring.
  • Complies with FDA Regulation CFR 21 and EU food contact material regulations.
  • Designed for areas requiring high levels of hygiene and effective water removal.