Learn More
MSC Carlisle Double Foam Squeegee, 18 inch, Blue
Catalog No. 502836722
Encompass
This double foam squeegee is 18 inches in length and comes in blue, designed for efficient and hygienic floor cleaning. Its one-piece construction minimizes potential contamination and makes it suitable for various surfaces, including delicate tile and marble floors. This versatile tool is compatible with standard threaded handles.
  • Safe to use on delicate tile and marble floors.
  • One-piece construction minimizes contamination.
  • Designed for use with standard threaded handles.
  • Universal thread adapter accepts both Euro or Acme threaded handles.
  • Available in multiple colors for color-coded cleaning systems.
  • Constructed with a durable polypropylene frame.
  • All plastic construction will not rust.
Catalog No. 50-283-6722 Supplier MSC Supplier No. 47167226
This double foam squeegee is 18 inches in length and comes in blue, designed for efficient and hygienic floor cleaning. Its one-piece construction minimizes potential contamination and makes it suitable for various surfaces, including delicate tile and marble floors. This versatile tool is compatible with standard threaded handles.
  • Safe to use on delicate tile and marble floors.
  • One-piece construction minimizes contamination.
  • Designed for use with standard threaded handles.
  • Universal thread adapter accepts both Euro or Acme threaded handles.
  • Available in multiple colors for color-coded cleaning systems.
  • Constructed with a durable polypropylene frame.
  • All plastic construction will not rust.