Learn More
Brady™ Double Bottle Eye Wash Station
Catalog No. 19129467 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Fluorescent Green
Catalog No. Color
19-129-467 Fluorescent Green
1 options
Catalog No. 19-129-467 Supplier Brady™ Supplier No. PD997E
  • $258.00
    $201.65 / Each of 1
    Save $56.35
    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Eye wash station with a drop down front panel that allows for quick access to eyewash bottles.
  • Proper usage instructions are conveniently silk-screened on the door
  • Station can be wall or table mounted
  • Holds two bottles of eye wash
Specifications
Color Fluorescent Green
Material Acrylic