Learn More
Accuflow Systems Double braid Reinforced Tubing
Supplier:  Accuflow Systems ONESILDB10001500
Encompass_Preferred
Purity One™ OneSil double-braid reinforced biopharmaceutical grade silicone tubing has been formulated for demanding high-pressure applications in biopharmaceutical processing. This flexible silicone tubing is ideal for use with radially crimped stainless-steel fittings and has higher pressure ratings than non-reinforced tubing, making it an excelling choice for bulk transfer, SIP/CIP, filtration, and buffer and media prep. Full validation guide and USP <665> Extractables testing available.
Catalog No. 50-299-5702