Learn More
Cole-Parmer Cole-Parmer Double Buret Clamp, Nickel-Plated Zinc, with Support Stand and Rod; Holds Up to 100 ml Burets
Catalog No. NC1499725 Shop All Cole-Parmer Products
Encompass_Preferred
Hold one or two burettes up to 100mL. The one-handed, scissor-like mechanism provides quick and easy adjustment. Clamp is available separately or together with stand. Choose from clamp only double burette or clamp with support stand and rod.
Catalog No. NC1499725 Supplier Cole-Parmer Supplier No. 0802260
Hold one or two burettes up to 100mL. The one-handed, scissor-like mechanism provides quick and easy adjustment. Clamp is available separately or together with stand. Choose from clamp only double burette or clamp with support stand and rod.