Learn More
Electron Microscopy Sciences Double Clamp with Stand 1 each
SDP
Catalog No. 50996767
Encompass_Preferred
Two adjustable arms with alligator jaws in a heavy base. A versatile tool which can be adjusted to any position and firmly hold small items of any shape.
Catalog No. 50-996-767 Supplier Electron Microscopy Sciences Supplier No. 6212301
Two adjustable arms with alligator jaws in a heavy base. A versatile tool which can be adjusted to any position and firmly hold small items of any shape.