Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Double Coated Film Tape

Catalog No. 19072118 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
36 yd. x 2 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-118 36 yd. x 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-118 Supplier 3M™ Supplier No. 7000048621
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Double Coated Film Tape features a high-density polyethylene carrier with synthetic rubber adhesive on a densified kraft paper liner.

  • Combines high initial adhesion and good holding power to ensure it bonds to a variety of materials.
  • Paper liner has been specially treated to improve its strength and removability characteristics.
  • High density polyethylene carrier provides dimensional stability and tears by hand
  • Adhesive removes cleanly from many surfaces to ensure a relatively easy clean-up process
  • Excellent quick stick and high adhesion ensure a reliable bond to a variety of substrates
  • This tape is in compliance with the FDA 21 CFR 175.105 guidelines for indirect food contact
TRUSTED_SUSTAINABILITY

Specifications

Product Type Film Tape
For Use With (Application) Automotive, Commercial Solutions, Electronics, Manufacturing, Transportation
Color White
Dimensions (L x W) 36 yd. x 2 in.
Model 9589
Quantity 24 Rolls/Cs.
Thickness (English) 9 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.