Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Double Coated Paper Tape

Catalog No. 19072033 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
36 yd. x 1 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-033 36 yd. x 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-033 Supplier 3M™ Supplier No. 7000144721
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Double Coated Paper Tape is a hand-tearable double coated tape with a natural rubber adhesive system designed to offer excellent adhesion to many types of substrates.

  • Adhesive is mounted on a paper carrier treated to provide increased strength and removability characteristics from high internal strength surfaces.
  • 9 mil double coated paper tape of natural rubber adhesive with a paper release liner
  • 3M™ Adhesive 860 offers high initial adhesion to a variety of substrates
  • Heavy paper carrier is treated to improve strength and provide dimensional stability
  • Double coated tape is ideal for temporary attaching or fixturing
  • Solvent-free manufacturing process helps meet environmental best practices goals
TRUSTED_SUSTAINABILITY

Specifications

Product Type Paper Tape
For Use With (Application) Automotive, Commericial Solutions, Electronics, Manufacturing, Transportation
Color Natural
Dimensions (L x W) 36 yd. x 1 in.
Model 401M
Quantity 36 Rolls/Cs.
Thickness (English) 9 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.