Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Double Coated Tape

Catalog No. 19072091 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
36 yd. x 1/2 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-072-091 36 yd. x 1/2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-072-091 Supplier 3M™ Supplier No. 7000048428
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Double Coated Tape features 3M™ Adhesive 300, a medium-firm acrylic adhesive ideal for joining materials that are smooth, thin and have low residual stress.

  • Excellent for foam and gasket attachment, HSE and LSE plastics such as polyethylene
  • Quick flowing adhesive delivers high initial adhesion for faster lamination
  • 0.5 mil PET film carrier adds dimensional stability and easier handling during slitting and die-cutting
  • 3M™ Adhesive 300 aggressively bonds to many surfaces, including plastics like polyethylene
  • 55# 3.2 mil densified kraft liner makes for easy release
  • Adhesive is applied to both sides of a 0.5 mil PET film which adds structural stability
TRUSTED_SUSTAINABILITY

Specifications

Product Type Double Coated Tape
For Use With (Application) Automotive, Commericial Solutions, Electronics, Manufacturing, Mining, Oil and Gas, Transportation
Color Clear
Dimensions (L x W) 36 yd. x 1/2 in.
Model 444
Quantity 72 Rolls/Cs.
Thickness (English) 3.9 mil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.