Learn More
Grainger Double Coupling SS 2 in Pipe Size Socket
Catalog No. 502749396
Encompass_Preferred
The BLUCHER Double Coupling Stainless is a double coupling made of stainless steel. It has a 2 inch nominal pipe size and is designed for use with stainless steel drainage systems. The coupling has no hub ends and provides a secure joint connection. It can accommodate minor degrees of misalignment. The double coupling allows joining two pipes of the same nominal size. It is manufactured in the United States.
Catalog No. 50-274-9396 Supplier Grainger Supplier No. 14N998
The BLUCHER Double Coupling Stainless is a double coupling made of stainless steel. It has a 2 inch nominal pipe size and is designed for use with stainless steel drainage systems. The coupling has no hub ends and provides a secure joint connection. It can accommodate minor degrees of misalignment. The double coupling allows joining two pipes of the same nominal size. It is manufactured in the United States.