Learn More
Grainger Double Coupling SS 3 in Pipe Size Socket
Catalog No. 502751636
Encompass_Preferred
The BLUCHER Double Coupling Stainless is a double coupling made of stainless steel. It has a 2 inch nominal pipe size and is compatible with schedule 40 pipe. The coupling has a pressure rating of 232 PSI at 70°F. It is made from 316 stainless steel for corrosion resistance. The coupling has a polished finish and rubber sleeve gaskets for a watertight seal. It can be used to join and align pipes in process piping applications. The dimensions are 4.5 x 2.75 x 2.5 inches.
Catalog No. 50-275-1636 Supplier Grainger Supplier No. 14N999
The BLUCHER Double Coupling Stainless is a double coupling made of stainless steel. It has a 2 inch nominal pipe size and is compatible with schedule 40 pipe. The coupling has a pressure rating of 232 PSI at 70°F. It is made from 316 stainless steel for corrosion resistance. The coupling has a polished finish and rubber sleeve gaskets for a watertight seal. It can be used to join and align pipes in process piping applications. The dimensions are 4.5 x 2.75 x 2.5 inches.