Learn More
Alkali Scientific ASI™ Double Decker™ PCR Freezer Boxes, Hinged Lid, 5/Pack
SDP
Catalog No. 503026812
Encompass_Preferred
Double Decker™ PCR Freezer Box, Cardboard, Hinged Lid, 5.25×5.25×2in, FB2392,(R2392)392 place (two levels of 196 places - 14×14), for 0.1 and 0.2mL PCR tubes, strips, and plates This unique new freezer box has been designed to maximize storage capacity for PCR tubes.  With 196 wells on each level, it can accommodate 392 individual 0.1mL or 0.2mL tubes. For strip tubes, the box handles 40 strips of 8 or 28 strips of 12. The top level is a cardboard tray with two finger grips that allow easy access to the lower level. The two level system doubles the capacity while keeping the total box height under 2 inches (51mm) and assuring a good fit in standard height stainless steel racks. Fits 0.2mL / 0.1mL PCR consumables Tubes, strips, or plates Two levels of 14 × 14 (196) wells Max capacity: 392 × PCR Tubes Standard dimensions (51/4 × 2 inch) Hinged lid stays attached Drain slots on bottom
Catalog No. 50-302-6812 Supplier Alkali Scientific Supplier No. FB2392
Double Decker™ PCR Freezer Box, Cardboard, Hinged Lid, 5.25×5.25×2in, FB2392,(R2392)392 place (two levels of 196 places - 14×14), for 0.1 and 0.2mL PCR tubes, strips, and plates This unique new freezer box has been designed to maximize storage capacity for PCR tubes.  With 196 wells on each level, it can accommodate 392 individual 0.1mL or 0.2mL tubes. For strip tubes, the box handles 40 strips of 8 or 28 strips of 12. The top level is a cardboard tray with two finger grips that allow easy access to the lower level. The two level system doubles the capacity while keeping the total box height under 2 inches (51mm) and assuring a good fit in standard height stainless steel racks. Fits 0.2mL / 0.1mL PCR consumables Tubes, strips, or plates Two levels of 14 × 14 (196) wells Max capacity: 392 × PCR Tubes Standard dimensions (51/4 × 2 inch) Hinged lid stays attached Drain slots on bottom