Learn More
AVANTOR MASTERFLEX (VWR LLC) Continuous-Duty Double-Diaphragm Pump; PP/Santoprene, 3.5 GPM, 115 VAC
Catalog No. NC1673296
Encompass
Multi-chambered pump offers efficient transfer of mild acids, bases, and light oils. Thermal overload protected, permanent-magnet motor. Pump includes 6-ft power cord with three-prong plug. Order replacement diaphragms or valve kits under accessories.
Catalog No. NC1673296 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7510200
Multi-chambered pump offers efficient transfer of mild acids, bases, and light oils. Thermal overload protected, permanent-magnet motor. Pump includes 6-ft power cord with three-prong plug. Order replacement diaphragms or valve kits under accessories.