Learn More
Braintree Scientific Double Edge Blades 0.1mm For 0.5mm Matrice 10Pk
SDP
Catalog No. 501954414
Encompass
Stainless Steel Brain Matrices These highly polished high quality stainless steel matrices are precisely machined to ensure reproducible sections. The brain matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1mm or 0.5mm intervals. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4414 Supplier Braintree Scientific Supplier No. BSSS05
Stainless Steel Brain Matrices These highly polished high quality stainless steel matrices are precisely machined to ensure reproducible sections. The brain matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1mm or 0.5mm intervals. **Sold for research purposes only and not for human use**