Learn More
Ted Pella Inc Double Edge Stainless Steel Cutting Blades, 250/box
SDP
Catalog No. NC1644135
Encompass
PTFE-Coated stainless steel, individually wrapped and bulk packed. Break in half lengthwise for use (protect fingers and use eye protection).Suitable blade for PELCO easiSlicer™, Microslicer™, Vibratome and other brands of vibratory microtomes. Suitable alternative to Feather blades when used as is (unbroken) in McIlwain Tissue Chopper.
Catalog No. NC1644135 Supplier Ted Pella Inc Supplier No. 1216
PTFE-Coated stainless steel, individually wrapped and bulk packed. Break in half lengthwise for use (protect fingers and use eye protection).Suitable blade for PELCO easiSlicer™, Microslicer™, Vibratome and other brands of vibratory microtomes. Suitable alternative to Feather blades when used as is (unbroken) in McIlwain Tissue Chopper.