Learn More
Med Vet International Double End Elevator Small 2MM, J0042W
Catalog No. 5000585132
Encompass_Preferred
This double-ended elevator has a unique veterinary design. The fine tips are both curved with an elevator grove. One end of the groove is on the inside concave side and the other end of the groove is on the outside convex side. This allows the elevator to follow the large root curves which exist on many animal teeth (i.e., deciduous canine teeth). Stainless steel knurled 71/4"L handle. Feline. 2mm.
Catalog No. 50-005-85132 Supplier Med Vet International Supplier No. JOR42W
This double-ended elevator has a unique veterinary design. The fine tips are both curved with an elevator grove. One end of the groove is on the inside concave side and the other end of the groove is on the outside convex side. This allows the elevator to follow the large root curves which exist on many animal teeth (i.e., deciduous canine teeth). Stainless steel knurled 71/4"L handle. Feline. 2mm.