Learn More
Electron Microscopy Sciences Double End Seeker/Probe
SDP
Catalog No. 5028542
Encompass_Preferred
Single end stainless steel probe with a hexagonal handle which comes with one fine point curved tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip.
Catalog No. 50-285-42 Supplier Electron Microscopy Sciences Supplier No. 7294502
Single end stainless steel probe with a hexagonal handle which comes with one fine point curved tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip. Double ended probe which comes with one end tapered to a point and one end curved with a rounded tip.